1-Boc-3-(ethylamino)azetidine 95%

Lieferant: Thermo Scientific
Achtung

H57941.03
H57941.03EA 117 EUR
H57941.03
1-Boc-3-(ethylamino)azetidine 95%
1-Boc-3-(ethylamino)azetidine

Purity: 95% CAS No.: 454703-23-2 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 MDL No.: MFCD11520619

Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11520619
CAS-Nummer: 454703-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR