1-Boc-3-(methylamino)azetidine 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
tert-butyl 3-(methylamino)azetidine-1-carboxylate
H57951.03
H57951.03EA
158
EUR
H57951.03
1-Boc-3-(methylamino)azetidine 95%
1-Boc-3-(methylamino)azetidine
Purity: 95%
CAS No.: 454703-20-9
Molecular Formula: C9H18N2O2
Formula Weight: 186.25
MDL No.: MFCD08752496
Formel:
C₉H₁₈N₂O₂ Molecular Weight: 186,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08752496 CAS-Nummer: 454703-20-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...