3-Hydroxy-2-pyrrolidinone 95%

Lieferant: Thermo Scientific
Achtung

H57961.03
H57961.03EA 188 EUR
H57961.03
3-Hydroxy-2-pyrrolidinone 95%
3-Hydroxy-2-pyrrolidinone

Purity: 95% CAS No.: 15116-68-4 Molecular Formula: C4H7NO2 Formula Weight: 101.11 Storage Temperature: 2°C to 8°C Melting Point: 102-107° MDL No.: MFCD09751256

Formel: C₄H₇NO₂
Molecular Weight: 101,11 g/mol
Schmelzpunkt: 102…107 °C
MDL: MFCD09751256
CAS-Nummer: 15166-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR