4-(Maleimidomethyl)benzonitrile 97
Lieferant: Thermo Fisher Scientific
|
|
H58015.03
H58015.03EA
136
EUR
H58015.03
4-(Maleimidomethyl)benzonitrile 97
4-(Maleimidomethyl)benzonitrile
Purity: 97%
Molecular Formula: C12H8N2O2
Formula Weight: 212.20
UN No.: UN3439
MDL No.: MFCD16665080
Formel:
C₁₂H₈N₂O₂ |
MDL:
MFCD16665080 CAS-Nummer: 1249094-81-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...