2,4-Difluorobenzylzinc bromide 0.5 M in THF

Lieferant: Thermo Fisher Scientific
Gefahr


verpackt unter Argon in wiederversiegelbaren ChemSeal™ Flaschen

H58036.AD
H58036.ADEA 256 EUR
H58036.AD
2,4-Difluorobenzylzinc bromide 0.5 M in THF
2,4-Difluorobenzylzinc bromide

Packaged under Argon in resealable ChemSeal™ bottles


CAS No.: 307496-26-0 Molecular Formula: C7H5BrF2Zn Formula Weight: 272.40 Storage Temperature: 2°C to 8°C UN No.: UN3399 MDL No.: MFCD01311423

Formel: C₇H₅BrF₂Zn
Dichte: 0,995
Lagertemperatur: Kühlschrank
MDL: MFCD01311423
CAS-Nummer: 307496-26-0
UN: 3399
ADR: 4.3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR