Ethyl-2-amino-4-(4-pyridyl)thiophene-3-carboxylate 97%

Lieferant: Thermo Scientific
Achtung

H58069.03
H58069.03EA 161 EUR
H58069.03
Ethyl-2-amino-4-(4-pyridyl)thiophene-3-carboxylate 97%
Ethyl-2-amino-4-(4-pyridyl)thiophene-3-carboxylate

Purity: 97% CAS No.: 117516-88-8 Molecular Formula: C12H12N2O2S Formula Weight: 248.29 MDL No.: MFCD01922021

Formel: C₁₂H₁₂N₂O₂S
Molecular Weight: 248,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01922021
CAS-Nummer: 117516-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR