3-(Boc-aminomethyl)pyridine 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: tert-butyl (pyridin-3-ylmethyl)carbamate

H58183.06
H58183.06EA 72.5 EUR
H58183.06
3-(Boc-aminomethyl)pyridine 97%
3-(Boc-aminomethyl)pyridine

Purity: 97% CAS No.: 102297-41-6 Molecular Formula: C11H16N2O2 Formula Weight: 208.26 Flash Point: 22°(72°F) Refractive Index: 1.5080 Boiling Point: 130° UN No.: UN1993 MDL No.: MFCD06411296

Formel: C₁₁H₁₆N₂O₂
Siedepunkt: 130 °C
Flammpunkt: 22 °C (72 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD06411296
CAS-Nummer: 102297-41-6
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR