4H-Thieno[3,2-b]pyrrole-5-carboxylic acid 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
DAO-IN-2
H58200.03
H58200.03EA
174
EUR
H58200.03
4H-Thieno[3,2-b]pyrrole-5-carboxylic acid 97%
4H-Thieno[3,2-b]pyrrole-5-carboxylic acid
Purity: 97%
CAS No.: 39793-31-2
Molecular Formula: C7H5NO2S
Formula Weight: 167.19
Melting Point: 180°
MDL No.: MFCD07368542
Formel:
C₇H₅NO₂S Molecular Weight: 167,19 g/mol Schmelzpunkt: 180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368542 CAS-Nummer: 39793-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...