3-Chromanecarboxylic acid 97%

Lieferant: Thermo Fisher Scientific
Achtung

H58448.03
H58448.03EA 89.1 EUR
H58448.03
3-Chromanecarboxylic acid 97%
3-Chromanecarboxylic acid

Purity: 97% CAS No.: 115822-57-6 Molecular Formula: C10H10O3 Formula Weight: 178.18 Melting Point: 117-121° MDL No.: MFCD03783554

Formel: C₁₀H₁₀O₃
Schmelzpunkt: 117…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03783554
CAS-Nummer: 115822-57-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR