4-(Ethoxycarbonyl)phenylzinc iodide 0.5 M in THF
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
verpackt unter Argon in wiederversiegelbaren ChemSeal™ Flaschen
H58484.AD
H58484.ADEA
237
EUR
H58484.AD
4-(Ethoxycarbonyl)phenylzinc iodide 0.5 M in THF
4-(Ethoxycarbonyl)phenylzinc iodide
Packaged under Argon in resealable ChemSeal™ bottles
CAS No.: 131379-16-3
Molecular Formula: C9H9IO2Zn
Formula Weight: 341.45
Storage Temperature: 2°C to 8°C
UN No.: UN3399
MDL No.: MFCD05664208
Formel:
C₉H₉IO₂Zn Molecular Weight: 341.453 g/mol Dichte: 1.023 Lagertemperatur: Kühlschrank |
MDL:
MFCD05664208 CAS-Nummer: 131379-16-3 UN: 3399 ADR: 4.3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...