2-Amino-5-chlor-6-methylpyridin 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
5-Chloro-6-methylpyridin-2-amine
H58501.06
H58501.06EA
362
EUR
H58501.06
2-Amino-5-chlor-6-methylpyridin 97%
2-Amino-5-chloro-6-methylpyridine
Purity: 97%
CAS No.: 36936-23-9
Molecular Formula: C6H7ClN2
Formula Weight: 142.59
MDL No.: MFCD09453335
Formel:
C₆H₇ClN₂ Schmelzpunkt: 70…75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09453335 CAS-Nummer: 36936-23-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...