2-Adamantylzinc bromide solution 0.5 M in THF
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
verpackt unter Argon in wiederversiegelbaren ChemSeal™ Flaschen
Packaged under Argon in resealable ChemSeal™ bottles
CAS No.: 171860-65-4
Molecular Formula: C10H15BrZn
Formula Weight: 280.51
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Density: 1.018
Sensitivity: Air Sensitive
Appearance: Yellow to brown to black
UN No.: UN3399
MDL No.: MFCD01311401
Formel:
C₁₀H₁₅BrZn Dichte: 1,018 Lagertemperatur: Kühlschrank |
MDL:
MFCD01311401 CAS-Nummer: 171860-65-4 UN: 3399 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...