6-Chloro-1H-benzoimidazole-2-carboxylic acid 97%

Lieferant: Thermo Fisher Scientific
Achtung

H58648.03
H58648.03EA 206 EUR
H58648.03
6-Chloro-1H-benzoimidazole-2-carboxylic acid 97%
6-Chloro-1H-benzoimidazole-2-carboxylic acid

Purity: 97% CAS No.: 39811-14-8 Molecular Formula: C8H5ClN2O2 Formula Weight: 196.59 MDL No.: MFCD06739053

Formel: C₈H₅ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06739053
CAS-Nummer: 39811-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR