3-Pyridin-4-ylpropionic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H58655.03
H58655.06
H58655.03EA
148
EUR
H58655.03
H58655.06
3-Pyridin-4-ylpropionic acid 97%
3-Pyridin-4-ylpropionic acid
Purity: 97%
CAS No.: 6318-43-0
Molecular Formula: C8H9NO2
Formula Weight: 151.16
Melting Point: 231°
Beilstein Reference: 122234
MDL No.: MFCD00084846
Formel:
C₈H₉NO₂ Schmelzpunkt: 231 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084846 CAS-Nummer: 6318-43-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...