2-Bromobenzylzinc bromide 0.5 M THF
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Argon_Chemseal
H58678.AD
H58678.ADEA
165
EUR
H58678.AD
2-Bromobenzylzinc bromide 0.5 M THF
2-Bromobenzylzinc bromide
Packaged under Argon in resealable ChemSeal™ bottles
CAS No.: 307496-27-1
Molecular Formula: C7H6Br2Zn
Formula Weight: 315.31
Storage Temperature: 2°C to 8°C
UN No.: UN3399
MDL No.: MFCD01311433
Formel:
C₇H₆Br₂Zn Molecular Weight: 315.313 g/mol Dichte: 1.015 Lagertemperatur: Kühlschrank |
MDL:
MFCD01311433 CAS-Nummer: 307496-27-1 UN: 3399 ADR: 4.3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...