4-(1-Hydroxyethyl)pyridine 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H58711.06
H58711.15
H58711.06EA
105
EUR
H58711.06
H58711.15
4-(1-Hydroxyethyl)pyridine 97%
4-(1-Hydroxyethyl)pyridine
Purity: 97%
CAS No.: 42732-22-9
Molecular Formula: C7H9NO
Formula Weight: 123.15
Storage Temperature: 2°C to 8°C
Melting Point: 51-55°
MDL No.: MFCD00077867
Formel:
C₇H₉NO Schmelzpunkt: 51…55 °C |
MDL:
MFCD00077867 CAS-Nummer: 23389-75-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...