2-Aminoimidazo[1,2-a]pyridin 97%

Lieferant: Thermo Fisher Scientific
Achtung

H58726.03
H58726.03EA 273 EUR
H58726.03
2-Aminoimidazo[1,2-a]pyridin 97%
2-Aminoimidazo[1,2-a]pyridin

Purity: 97% CAS No.: 28036-33-1 Molecular Formula: C7H7N3 Formula Weight: 133.15 Melting Point: 114-126° MDL No.: MFCD06660421

Formel: C₇H₇N₃
Schmelzpunkt: 114…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09836159
CAS-Nummer: 39588-26-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR