4,5-Dicyan-2-phenylimidazol ≥97%

Lieferant: Thermo Scientific
Achtung

H58800.04
H58800.04EA 120 EUR
H58800.04
4,5-Dicyan-2-phenylimidazol ≥97%
4,5-Dicyano-2-phenylimidazole
Formel: C₁₁H₆N₄
Molecular Weight: 194,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863325
CAS-Nummer: 50847-06-8
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR