5-(5-Methyl-2-furyl)thieno[2,3-d]pyrimidin-4(3H)-one 97%
Lieferant: Thermo Scientific
|
Achtung
|
H58812.06
H58812.06EA
1232
EUR
H58812.06
5-(5-Methyl-2-furyl)thieno[2,3-d]pyrimidin-4(3H)-one 97%
5-(5-Methyl-2-furyl)thieno[2,3-d]pyrimidin-4(3H)-one
Purity: 97%
Molecular Formula: C11H8N2O2S
Formula Weight: 232.25
MDL No.: MFCD22988976
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...