5-(5-Methyl-2-thienyl)thieno[2,3-d]pyrimidin-4-one 97%

Lieferant: Thermo Fisher Scientific
Achtung

H58856.03
H58856.03EA 353 EUR
H58856.03
5-(5-Methyl-2-thienyl)thieno[2,3-d]pyrimidin-4-one 97%
5-(5-Methyl-2-thienyl)thieno[2,3-d]pyrimidin-4-one

Purity: 97% Molecular Formula: C11H8N2OS2 Formula Weight: 248.31 MDL No.: MFCD06671220

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR