N-Boc-4-bromo-3-fluoroaniline 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: tert-Butyl N-(4-bromo-3-fluorophenyl)carbamate, tert-Butyl (4-bromo-3-fluorophenyl)carbamate

H59398.03
H59398.03EA 279 EUR
H59398.03
N-Boc-4-bromo-3-fluoroaniline 96%
N-Boc-4-bromo-3-fluoroaniline

Purity: 96% CAS No.: 868735-43-7 Molecular Formula: C11H13BrFNO2 Formula Weight: 290.13 Melting Point: 68-70° Beilstein Reference: 14460051 UN No.: UN2811 MDL No.: MFCD11976284

Formel: C₁₁H₁₃BrFNO₂
Molecular Weight: 290.132 g/mol
Schmelzpunkt: 68…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11976284
CAS-Nummer: 868735-43-7
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR