1-Ethyl-3-methylimidazoliumiodid ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-ethyl-3-methylimidazolium iodide
H59404.06
H59404.18
H59404.06EA
36.1
EUR
H59404.06
H59404.18
1-Ethyl-3-methylimidazoliumiodid ≥97%
1-Ethyl-3-methylimidazolium iodide
Formel:
C₆H₁₁IN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03701101 CAS-Nummer: 35935-34-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...