1-Ethyl-3-methylimidazoliummethylsulfat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-Ethyl-3-methyl-1H-imidazol-3-ium methyl sulphate

H59480.06 H59480.18
H59480.06EA 35.6 EUR
H59480.06 H59480.18
1-Ethyl-3-methylimidazoliummethylsulfat ≥98%
1-Ethyl-3-methylimidazolium methylsulphate
Formel: C₇H₁₄N₂O₄S
Dichte: 1,28 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD08457697
CAS-Nummer: 516474-01-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR