L(+)-Methyl-tert-leucinat Hydrochlorid ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: L-tert-Leucine methyl ester hydrochloride, H-Tle-OMe·HCl, H-Tle-OMe.HCl

H59563.03
H59563.03EA 220 EUR
H59563.03
L(+)-Methyl-tert-leucinat Hydrochlorid ≥97%
L(+)-Methyl-tert-leucinat Hydrochlorid
Formel: C₇H₁₅NO₂·HCl
Molecular Weight: 181.7 g/mol
Schmelzpunkt: 183 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00040604
CAS-Nummer: 63038-27-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR