4-Amino-2-fluorphenylboronsäurepinakolester 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Amino-2-fluorobenzeneboronic acid pinacol ester, 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
H59916.03
H59916.03EA
241
EUR
H59916.03
4-Amino-2-fluorphenylboronsäurepinakolester 96%
4-Amino-2-fluorphenylboronsäurepinakolester
Formel:
C₁₂H₁₇BFNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09951877 CAS-Nummer: 819057-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...