1,3,5-Tris-(4-carboxyphenyl)-benzol 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5'-(4-Carboxyphenyl)-[1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid

H60087.06
H60087.06EA 368 EUR
H60087.06
1,3,5-Tris-(4-carboxyphenyl)-benzol 97%
1,3,5-Tris(4-carboxyphenyl)benzene

Purity: 97% CAS No.: 50446-44-1 Molecular Formula: C27H18O6 Formula Weight: 438.43 Physical Form: Solid Melting Point: 322-327° UN No.: UN3077 MDL No.: MFCD10000888

Formel: C₂₇H₁₈O₆
Molecular Weight: 438,44 g/mol
Schmelzpunkt: 322…327 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD10000888
CAS-Nummer: 50446-44-1
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR