Oleanolic acid 97%

Lieferant: Thermo Fisher Scientific
Achtung

H60445.MD
H60445.MDEA 176 EUR
H60445.MD
Oleanolic acid 97%
Oleanolic acid
Formel: C₃₀H₄₈O₃
Molecular Weight: 456.71 g/mol
Schmelzpunkt: 300 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00064914
CAS-Nummer: 508-02-1
EINECS: 208-081-6

Bestellen

Spezifikation Testergebnisse

Purity 97%
Assay (Titration) >96,0%
Form Solid
Storage Temperature 2°C to 8°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR