1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene ≥97%
Lieferant: Thermo Fisher Scientific
Synonyme:
(+)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)benzene, (S,S)-Me-DUPHOS, 1,2-Bis((2S,5S)-2,5-dimethylphospholan-1-yl)benzene
H60647.MD
H60647.03
H60647.MDEA
133
EUR
H60647.MD
H60647.03
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene ≥97%
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene
It is a DuPhos and BPE ligands which are highly efficient privileged ligands.
Formel:
C₁₈H₂₈P₂ Molecular Weight: 306.37 g/mol Schmelzpunkt: 66…76 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00142322 CAS-Nummer: 136735-95-0 |
Spezifikation Testergebnisse
Sensitivity | Air Sensitive |
Solubility | Insoluble in water |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...