4-Hydroxy-o-toluylsäure ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-Hydroxy-2-methylbenzoic acid, 4-Hydroxy-o-toluic acid

H60810.09
H60810.09EA 217 EUR
H60810.09
4-Hydroxy-o-toluylsäure ≥98%
4-Hydroxy-o-toluylsäure
Formel: C₈H₈O₃
Molecular Weight: 152,15 g/mol
Schmelzpunkt: 177…181 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02182261
CAS-Nummer: 578-39-2
EINECS: 209-422-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR