(±)-2-Hydroxyisovaleriansäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Hydroxy-3-methylbutanoic acid, 2-Hydroxy-3-methylbutyric acid
H60872.03
H60872.03EA
76.7
EUR
H60872.03
(±)-2-Hydroxyisovaleriansäure ≥98%
(±)-2-Hydroxyisovaleriansäure
Formel:
C₅H₁₀O₃ Molecular Weight: 118,13 g/mol Schmelzpunkt: 82…84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004242 CAS-Nummer: 4026-18-0 EINECS: 223-697-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...