4,5-Dihydroxyanthraquinone-2-carboxylic acid 98%

Lieferant: Thermo Fisher Scientific
Achtung

H60902.03
H60902.03EA 553 EUR
H60902.03
4,5-Dihydroxyanthraquinone-2-carboxylic acid 98%
4,5-Dihydroxyanthraquinone-2-carboxylic acid

Purity: 98% CAS No.: 478-43-3 Molecular Formula: C15H8O6 Formula Weight: 284.22 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: Dark yellow Melting Point: 300° Beilstein Reference: 2222155 MDL No.: MFCD00009618

Formel: C₁₅H₈O₆
Schmelzpunkt: >300 °C
MDL: MFCD00009618
CAS-Nummer: 478-43-3
EINECS: 207-521-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR