2,2-Dimethyl-4-pentensäure 95%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2,2-Dimethylpent-4-enoic acid

H60940.06
H60940.06EA 247 EUR
H60940.06
2,2-Dimethyl-4-pentensäure 95%
2,2-Dimethyl-4-pentenoic acid

Purity: 95% CAS No.: 16386-93-9 Molecular Formula: C7H12O2 Formula Weight: 128.17 Flash Point: 88°(190°F) Physical Form: Liquid Density: 0.933 Refractive Index: 1.431 Appearance: Yellow Boiling Point: 104-108° UN No.: UN3265 MDL No.: MFCD00012304

Formel: C₇H₁₂O₂
Molecular Weight: 128,17 g/mol
Siedepunkt: 104…108 °C
Dichte: 0,933
Flammpunkt: 88 °C (190 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012304
CAS-Nummer: 16386-93-9
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR