3-Amino-6-chlor-4-methylpyridin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3-Amino-6-chloro-4-picoline, 5-Amino-2-chloro-4-methylpyridine, 6-Chloro-4-methylpyridin-3-amine

H61115.14
H61115.14EA 207 EUR
H61115.14
3-Amino-6-chlor-4-methylpyridin 98%
3-Amino-6-chloro-4-methylpyridine
Formel: C₆H₇ClN₂
Molecular Weight: 142.59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03427661
CAS-Nummer: 66909-38-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR