4-Cyan-2-fluorphenylboronsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Cyano-2-fluorophenylboronic Acid, 4-Cyano-2-fluorobenzeneboronic acid

H61134.03
H61134.03EA 189 EUR
H61134.03
4-Cyan-2-fluorphenylboronsäure ≥98%
4-Cyan-2-fluorphenylboronsäure

Purity: 98% CAS No.: 1150114-77-4 Molecular Formula: C7H5BFNO2 Formula Weight: 164.93 Physical Form: Solid

Formel: C₇H₅BFNO₂
Molecular Weight: 164,93 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07644472
CAS-Nummer: 1150114-77-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR