5-Brom-2-chlornicotinsäure 96%

Lieferant: Thermo Scientific
Achtung

H61154.22
H61154.22EA 116 EUR
H61154.22
5-Brom-2-chlornicotinsäure 96%
5-Brom-2-chlornicotinsäure

Purity: 96% CAS No.: 29241-65-4 Molecular Formula: C6H3BrClNO2 Formula Weight: 236.45 Physical Form: Solid MDL No.: MFCD03844847

Formel: C₆H₃BrClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03844847
CAS-Nummer: 29241-65-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR