3-Brom-2-nitroanilin ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
H61169.06
H61169.06EA
360
EUR
H61169.06
3-Brom-2-nitroanilin ≥98%
3-Brom-2-nitroanilin
Purity: 98%
CAS No.: 7138-15-0
Molecular Formula: C6H5BrN2O2
Formula Weight: 217.02
Physical Form: Crystalline
Appearance: Dark red
UN No.: 2811
MDL No.: MFCD08063248
Formel:
C₆H₅BrN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08063248 CAS-Nummer: 7138-15-0 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...