1-(4-Cyanphenyl)-2,5-dimethylpyrrol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-(2,5-Dimethyl-1-pyrrolyl)benzonitrile
H61178.06
H61178.06EA
222
EUR
H61178.06
1-(4-Cyanphenyl)-2,5-dimethylpyrrol ≥98%
1-(4-Cyanophenyl)-2,5-dimethylpyrrole
Formel:
C₁₃H₁₂N₂ Molecular Weight: 196,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03228548 CAS-Nummer: 119516-86-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...