2-Morpholinoacetic acid hydrochloride 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Morpholinoacetic acid hydrochloride, Morpholin-4-ylacetic acid hydrochloride, 4-Morpholineacetic acid hydrochloride
H61188.06
H61188.06EA
275
EUR
H61188.06
2-Morpholinoacetic acid hydrochloride 95%
2-Morpholinoacetic acid hydrochloride
Purity: 95%
CAS No.: 89531-58-8
Molecular Formula: C6H11NO3.HCl
Formula Weight: 181.62
Physical Form: Powder
Appearance: White
Beilstein Reference: 3701710
MDL No.: MFCD03540888
Formel:
C₆H₁₂ClNO₃ Molecular Weight: 181,616 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03540888 CAS-Nummer: 89531-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...