3-Brom-5-fluoranilin 95%

Lieferant: Thermo Fisher Scientific
Achtung

H61238.03
H61238.03EA 127 EUR
H61238.03
3-Brom-5-fluoranilin 95%
3-Brom-5-fluoranilin

Purity: 95% CAS No.: 134168-97-1 Molecular Formula: C6H5BrFN Formula Weight: 190.01 Physical Form: Liquid Appearance: Brown UN No.: 2810 MDL No.: MFCD07779529

Formel: C₆H₅BrFN
Lagertemperatur: Raumtemperatur
MDL: MFCD07779529
CAS-Nummer: 134168-97-1
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR