2,6-Dichlor-3-methylpyridin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
H61271.03
H61271.03EA
102
EUR
H61271.03
2,6-Dichlor-3-methylpyridin ≥98%
2,6-Dichloro-3-methylpyridine
Formel:
C₆H₅Cl₂N Molecular Weight: 162,02 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00137847 CAS-Nummer: 58584-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...