2-Amino-3-nitro-5-fluorpyridin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H61293.06
H61293.06EA
527
EUR
H61293.06
2-Amino-3-nitro-5-fluorpyridin 98%
2-Amino-3-nitro-5-fluorpyridin
Formel:
C₅H₄FN₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972413 CAS-Nummer: 212268-12-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...