Methyl-6-fluornicotinat 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
6-Fluoronicotinic acid methyl ester, Methyl 6-fluoropyridine-3-carboxylate
H61302.03
H61302.03EA
121
EUR
H61302.03
Methyl-6-fluornicotinat 95%
Methyl-6-fluornicotinat
Formel:
C₇H₆FNO₂ Molecular Weight: 155,13 g/mol Schmelzpunkt: 48…52 °C Flammpunkt: 99 °C (210 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095098 CAS-Nummer: 1427-06-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...