1,3-Dichlor-2-fluor-5-nitrobenzol 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,5-Dichloro-4-fluoro-1-nitrobenzene
H61321.06
H61321.14
H61321.06EA
50.7
EUR
H61321.06
H61321.14
1,3-Dichlor-2-fluor-5-nitrobenzol 95%
1,3-Dichloro-2-fluoro-5-nitrobenzene
Formel:
C₆H₂Cl₂FNO₂ Molecular Weight: 209.99 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00129963 CAS-Nummer: 3107-19-5 EINECS: 221-466-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...