3-Benzo[b]thiophensulfonylchlorid 97%

Lieferant: Thermo Fisher Scientific
Gefahr

H61368.MD H61368.03
H61368.MDEA 76.4 EUR
H61368.MD H61368.03
3-Benzo[b]thiophensulfonylchlorid 97%
3-Benzo[b]thiophensulfonylchlorid

Purity: 97% CAS No.: 18494-87-6 Molecular Formula: C8H5ClO2S2 Formula Weight: 232.71 Storage Temperature: 2°C to 8°C Melting Point: 86-88° Beilstein Reference: 1285265 UN No.: UN3261 MDL No.: MFCD02681897

Formel: C₈H₅ClO₂S₂
Schmelzpunkt: 86…88 °C
MDL: MFCD02681897
CAS-Nummer: 18494-87-6
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR