Imidazo[1,2-b]pyridazin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H61377.06 H61377.14
H61377.06EA 32 EUR
H61377.06 H61377.14
Imidazo[1,2-b]pyridazin ≥98%
Imidazo[1,2-b]pyridazin
Formel: C₆H₅N₃
Molecular Weight: 119.12 g/mol
Schmelzpunkt: 54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07782103
CAS-Nummer: 766-55-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR