2-Brom-p-anisaldehyd ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Bromo-4-methoxybenzaldehyde
H61379.03
H61379.06
H61379.03EA
144
EUR
H61379.03
H61379.06
2-Brom-p-anisaldehyd ≥98%
2-Brom-p-anisaldehyd
Formel:
C₈H₇BrO₂ Molecular Weight: 215.04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09788636 CAS-Nummer: 43192-31-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...