2-Brom-p-anisaldehyd ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Bromo-4-methoxybenzaldehyde

H61379.03 H61379.06
H61379.03EA 144 EUR
H61379.03 H61379.06
2-Brom-p-anisaldehyd ≥98%
2-Brom-p-anisaldehyd
Formel: C₈H₇BrO₂
Molecular Weight: 215.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09788636
CAS-Nummer: 43192-31-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR