4-Brom-o-anisidin ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Methoxy-4-bromanilin, 2-Amino-5-bromanisol, 4-Brom-2-methoxyanilin

H61454.06
H61454.06EA 76.5 EUR
H61454.06
4-Brom-o-anisidin ≥98%
4-Brom-o-anisidin

Purity: 98% CAS No.: 59557-91-4 Molecular Formula: C7H8BrNO Formula Weight: 202.05 UN No.: 2811 MDL No.: MFCD01204266

Formel: C₇H₈BrNO
Molecular Weight: 202,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01204266
CAS-Nummer: 59557-91-4
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR