3-Amino-6-chlor-2-methylpyridin 95%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 6-Chloro-2-methylpyridin-3-amine

H61465.06
H61465.06EA 63 EUR
H61465.06
3-Amino-6-chlor-2-methylpyridin 95%
3-Amino-6-chloro-2-methylpyridine
Formel: C₆H₇ClN₂
Molecular Weight: 142,59 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD03095220
CAS-Nummer: 164666-68-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR