1-Brom-5-fluor-2-nitrobenzol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Brom-4-fluornitrobenzol, 1-Brom-3-fluor-6-nitrobenzol

H61466.06 H61466.14
H61466.06EA 53 EUR
H61466.06 H61466.14
1-Brom-5-fluor-2-nitrobenzol ≥98%
1-Brom-5-fluor-2-nitrobenzol
Formel: C₆H₃BrFNO₂
Molecular Weight: 220 g/mol
Schmelzpunkt: 41…45 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00792441
CAS-Nummer: 700-36-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR